Learn More
RABL2A Mouse anti-Human, Clone: OTI4A3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF502019
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the RAB gene family which belongs to the RAS GTPase superfamily. The proteins in the family of RAS-related signaling molecules are small GTP-binding proteins that play important roles in the regulation of exocytotic and endocytotic pathways. This gene maps to the site of an ancestral telomere fusion event and may be a subtelomeric gene. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| RABL2A | |
| Monoclonal | |
| Unconjugated | |
| RABL2A | |
| RAB, member of RAS oncogene family like 2A | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11159 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4A3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9UBK7 | |
| RABL2A | |
| Full length human recombit protein of human RABL2A produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.