Learn More
Novus Biologicals™ RABL2A Recombinant Protein Antigen
Product Code. 18184459 Shop All Bio Techne Products
Click to view available options
Protein Length:
DKTKPSELDQGKYDADDNVKIICLGDSAVGKSKLMERFLMDGFQPQQLSTYALTLYKHTATV
KLFNDAIRLAVSYKQNSQDFMDEIFQELENFSLEQEEEDVPDQEQSSSIETPS
Unit Size:
0.10mL
0.1mL
Product Code. 18184459
Brand: Novus Biologicals™ NBP246681PEP
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human RABL2A The RABL2A Recombinant Protein Antigen is derived from E. coli. The RABL2A Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY
Specifications
Protein Length KLFNDAIRLAVSYKQNSQDFMDEIFQELENFSLEQEEEDVPDQEQSSSIETPS
Purity >80% by SDS-PAGE and Coomassie blue staining
Common Name RABL2A Recombinant Protein Antigen
Content And Storage Store at −20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4
For Use With (Application) AC
Gene Alias FLJ78724, MGC117180
Gene Symbol RABL2A
Label Type Unlabeled
Product Type Recombinant Protein Antigen
Quantity 0.1 mL
Regulatory Status RUO
Source E.coli
Specific Reactivity Human