missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABL2B Monoclonal Antibody (1B10)

RABL2B Monoclonal Antibody (1B10)
Brand: Invitrogen MA521282
This item is not returnable.
View return policy
Description
Specifications
| RABL2B | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9UNT1 | |
| RABL2B | |
| RABL2B (AAH14879, 1 a.a. ~ 229 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 1B10 | |
| Unconjugated | |
| RABL2B | |
| HOX1B; RAB, member of RAS oncogene family-like 2B; RABL2B; rab-like protein 2B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11158 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction