missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RABL5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104052
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RABL5.
Small GTPase-like component of the intraflagellar transport (IFT) complex B.
Specifications
| RABL5 | |
| Polyclonal | |
| Unconjugated | |
| Ift22 | |
| 3110017O03Rik; AI835745; AI847427; IFT22; intraflagellar transport 22; intraflagellar transport 22 homolog; intraflagellar transport protein 22 homolog; RAB, member of RAS oncogene family-like 5; RAB, member RAS oncogene family-like 5; Rabl5; rab-like protein 5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 288585, 64792 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5FVJ7, Q9H7X7 | |
| Ift22 | |
| A synthesized peptide derived from human IFT22(Accession Q9H7X7), corresponding to amino acid residues E31-A81. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction