missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabphilin 3A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | Rabphilin 3A |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227260
|
Novus Biologicals
NBP3-38317-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30226744
|
Novus Biologicals
NBP3-38317-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rabphilin 3A Polyclonal antibody specifically detects Rabphilin 3A in Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| Rabphilin 3A | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Membrane Trafficking and Chaperones, Neuroscience, Phospho Specific | |
| PBS (pH 7.3), 50% glycerol | |
| 22895 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| Exophilin-1, KIAA0985exophilin-1, rabphilin, rabphilin 3A homolog (mouse), rabphilin-3A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human Rabphilin 3A (NP_055769.2).,, Sequence:, MTDTVFSNSSNRWMYPSDRPLQSKLQAGWSVHPGGQPDRQRKQEELTDEEKEIINRVIARAEKMEEMEQERIGRLVDRLENMRKNVAGDGVNRCILCGEQLGMLGSACVVCEDCKKNVCTKCGVETNNRLHSVWLCKICIEQREVWKRSGAWFFKGFPKQVLPQPMPIKKTKPQQPVSEPAAPEQPAPEPKHPARAPARGDSEDRRGPGQKTGPDPASAPGRGNYGPPVRRASEARMSSSSRDSESWDHSGGAGDSSRSPAGLRRANSVQASRPAPGSVQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title