Learn More
rac 1-Palmitoyl-3-chloropropanediol, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Palmitoyl-3-chloropropanediol |
| CAS | 30557-04-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19 H37 Cl O3 |
| Quantity | 50 mg |
| Synonym | Palmitic acid','3-chloro-2-hydroxypropyl ester (8CI), 3-Chloro-1,2-propanediol 1-palmitate, Rac-1-palmitoyl-3-chloropropane-1,2-diol |
| InChI Formula | InChI=1S/C19H37ClO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(22)23-17-18(21)16-20/h18,21H,2-17H2,1H3 |
| SMILES | CCCCCCCCCCCCCCCC(=O)OCC(O)CCl |
| IUPAC Name | (3-chloro-2-hydroxypropyl) hexadecanoate |
| Molecular Weight (g/mol) | 348.95 |
| Show More |