missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rac Biotin-d4, TRC
Specifications
Specifications
| Chemical Name or Material | (±)-Biotin-D4 (pentanoic acid 2,2,3,3-D4) |
| CAS | 1217850-77-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 D4 H12 N2 O3 S |
| Quantity | 0.5 mg |
| Synonym | rac Biotin-d4, (±)-Biotin-d4, rel-(3aR,4R,6aS)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanoic-2,2,3,3-d4 acid, 5-[(3aRS,4RS,6aSR)-2-Oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-2,2,3,3-tetradeuteriopentanoic acid, (±)-Biotin-D4 (pentanoic acid 2,2,3,3-D4) |
| InChI Formula | InChI=1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-/m0/s1/i1D2,2D2 |
| SMILES | [2H]C([2H])(C[C@@H]1SC[C@@H]2NC(=O)N[C@H]12)C([2H])([2H])CC(=O)O |
| IUPAC Name | 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-3,3,4,4-tetradeuterio-pentanoic acid |
| Molecular Weight (g/mol) | 248.34 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?