Learn More
rac-Keto Labetalol Hydrochloride (~90%), TRC
Specifications
Specifications
| Chemical Name or Material | rac-Keto Labetalol Hydrochloride |
| CAS | 96441-14-4 |
| Molecular Formula | C19H23ClN2O3 |
| Quantity | 100 mg |
| Synonym | Labetalone, 2-Hydroxy-5-[2-[(1-methyl-3-phenylpropyl)amino]acetyl]benzamide Hydrochloride, 2-Hydroxy-5-[[(1-methyl-3-phenylpropyl)amino]acetyl]benzamide Monohydrochloride, Labetalone Hydrochloride |
| InChI Formula | InChI=1S/C19H22N2O3.ClH/c1-13(7-8-14-5-3-2-4-6-14)21-12-18(23)15-9-10-17(22)16(11-15)19(20)24;/h2-6,9-11,13,21-22H,7-8,12H2,1H3,(H2,20,24);1H |
| SMILES | CC(NCC(C1=CC(C(N)=O)=C(O)C=C1)=O)CCC2=CC=CC=C2.Cl |
| IUPAC Name | 2-hydroxy-5-[2-(4-phenylbutan-2-ylamino)acetyl]benzamide;hydrochloride |
| Molecular Weight (g/mol) | 362.85 |
| Formula Weight | 362.1397 |