Learn More
rac-N-Desmethylselegiline Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal
Product Code. 30461508
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461508 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30461508 Supplier LoGiCal Supplier No. LGCAMP0216.0901
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
rac-N-Desmethylselegiline Hydrochloride 1.0 mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material rac-N-Desmethylselegiline Hydrochloride
CAS 2588-96-7
Analyte or Component Names rac-N-Desmethylselegiline Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C12 H15 N . Cl H
Certifications/Compliance ISO 17034
Synonym N-[(1 RS)-1-Methyl-2-phenylethyl]prop-2-yn-1-amine Hydrochloride, Selegiline Hydrochloride Imp. D (EP) as Racemate Hydrochloride, rac-Demethylselegiline Hydrochloride, rac-N-Desmethylselegiline Hydrochloride, Selegiline Hydrochloride Imp. D (EP), Selegiline Imp. D (EP), Selegiline Hydrochloride Impurity D as Racemate Hydrochloride, Selegiline Impurity D as Racemate Hydrochloride
InChI Formula InChI=1 S/C12H15N.ClH/c1-3-9-13-11(2)10-12-7-5-4-6-8-12;/h1,4-8,11,13 H,9-10H2,2H3;1 H
SMILES Cl.CC(Cc1ccccc1)NCC#C
IUPAC Name 1-phenyl-N-prop-2-ynylpropan-2-amine;hydrochloride
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 209.72
Quantity 1 mL
Formula Weight 209.0971 g/mol