missing translation for 'onlineSavingsMsg'
Learn More

rac O-Desmethyl Naproxen-d3, TRC

Product Code. 30367095
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367095 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30367095 Supplier Toronto Research Chemicals Supplier No. TRCD2921271MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac O-Desmethyl Naproxen-d3
CAS 1122399-99-8
Molecular Formula C13 D3 H9 O3
Quantity 1 mg
Synonym 2-Naphthaleneacetic acid, 6-hydroxy-α-(methyl-d3)- (ACI), 6-Hydroxy-α-(methyl-d3)-2-naphthaleneacetic acid (ACI), (±)-Naproxen-O-desmethyl-D3 (methyl-D3), (R,S)-Naproxen-O-desmethyl-d3, 6-O-Desmethylnaproxen, 2-(6-Hydroxy-2-naphthyl)propanoic-3,3,3-d3 acid, 2-(6-Hydroxynaphthalen-2-yl)-propanoic acid-3,3,3-d3, 6-O-Desmethyl naproxen-d3, (±)-O-Desmethylnaproxen-d3 (α-methyl-d3), rac O-Desmethyl Naproxen-d3
InChI Formula InChI=1S/C13H12O3/c1-8(13(15)16)9-2-3-11-7-12(14)5-4-10(11)6-9/h2-8,14H,1H3,(H,15,16)/i1D3
SMILES [2H]C([2H])([2H])C(C(=O)O)c1ccc2cc(O)ccc2c1
IUPAC Name 3,3,3-trideuterio-2-(6-hydroxynaphthalen-2-yl)propanoic acid
Molecular Weight (g/mol) 219.251
Formula Weight 219.097
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.