Learn More
(Rac)-Rotigotine hydrochloride, MedChemExpress
Product Code. 16496746
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16496746
Brand: Medchem Express HY15394
MedChemExpress (Rac)-Rotigotine hydrochloride is a racemate of Rotigotine. Rotigotine is a full agonist of dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor, with Kis of 0.71 nM, 4-15 nM, and 83 nM for the dopamine D3 receptor and D2, D5, D4 receptors, and dopamine D1 receptor.
(Rac)-Rotigotine hydrochloride is a racemate of Rotigotine. Rotigotine is a full agonist of dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor, with Kis of 0.71 nM, 4-15 nM, and 83 nM for the dopamine D3 receptor and D2, D5, D4 receptors, and dopamine D1 receptor.
Specifications
Chemical Name or Material (Rac)-Rotigotine hydrochloride
CAS 102120-99-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H26ClNOS
Quantity 100 mg
Solubility Information DMSO : ≥ 50 mg/mL (142.07 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES OC1=C2CCC(N(CCC)CCC3=CC=CS3)CC2=CC=C1.[H]Cl
Molecular Weight (g/mol) 351.93
Formula Weight 351.93
Percent Purity 98.66%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only