missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Rac)-Rotigotine hydrochloride, MedChemExpress
Description
(Rac)-Rotigotine hydrochloride is a racemate of Rotigotine. Rotigotine is a full agonist of dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor, with Kis of 0.71 nM, 4-15 nM, and 83 nM for the dopamine D3 receptor and D2, D5, D4 receptors, and dopamine D1 receptor.
Specifications
Specifications
| Chemical Name or Material | (Rac)-Rotigotine hydrochloride |
| CAS | 102120-99-0 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C19H26ClNOS |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 50 mg/mL (142.07 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | OC1=C2CCC(N(CCC)CCC3=CC=CS3)CC2=CC=C1.[H]Cl |
| Molecular Weight (g/mol) | 351.93 |
| Formula Weight | 351.93 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?