Learn More
rac Tenofovir-d6, TRC
Specifications
Specifications
| Chemical Name or Material | Tenofovir-D6 |
| CAS | 1020719-94-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C9 2H6 H8 N5 O4 P |
| Quantity | 10 mg |
| Synonym | P-[[1-[(6-amino-9H-purin-9-yl)methyl-d2]ethoxy-1,2,2,2-d4]methyl]phosphonic acid |
| InChI Formula | InChI=1S/C9H14N5O4P/c1-6(18-5-19(15,16)17)2-14-4-13-7-8(10)11-3-12-9(7)14/h3-4,6H,2,5H2,1H3,(H2,10,11,12)(H2,15,16,17)/i1D3,2D2,6D |
| SMILES | [2H]C([2H])([2H])C([2H])(OCP(=O)(O)O)C([2H])([2H])n1cnc2c(N)ncnc12 |
| IUPAC Name | [1-(6-aminopurin-9-yl)-1,1,2,3,3,3-hexadeuteriopropan-2-yl]oxymethylphosphonic acid |
| Molecular Weight (g/mol) | 293.25 |
| Show More |