missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rac1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99370-50ul
This item is not returnable.
View return policy
Description
Rac1 Polyclonal antibody specifically detects Rac1 in Human samples. It is validated for ELISA
Specifications
| Rac1 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| Cell migration-inducing gene 5 protein, MGC111543, p21-Rac1migration-inducing gene 5, Ras-like protein TC25, ras-related C3 botulinum toxin substrate 1, ras-related C3 botulinum toxin substrate 1 (rho family, small GTP bindingprotein Rac1), rho family, small GTP binding protein Rac1, TC25, TC-25 | |
| Produced in rabbits immunized with purified, recombinant Human Rac1 (Accession#: NP_008839.2; Met1-Cys189) | |
| 50 μg | |
| Breast Cancer, Cancer, mTOR Pathway | |
| 5879 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction