missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAC3 Antibody (JE56-48), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32888
This item is not returnable.
View return policy
Description
RAC3 Monoclonal antibody specifically detects RAC3 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| RAC3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000 | |
| p21-Rac3, ras-related C3 botulinum toxin substrate 3, ras-related C3 botulinum toxin substrate 3 (rho family, small GTP bindingprotein Rac3), rho family, small GTP binding protein Rac3 | |
| Recombinant protein within Human RAC3 aa 71-170/ 192. (Uniprot: P60763) | |
| 100 μL | |
| Signal Transduction | |
| 5881 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE56-48 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction