missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAC3 Antibody (S01-6I8), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 444.00 - € 680.00
Specifications
| Antigen | RAC3 |
|---|---|
| Clone | S01-6I8 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18677523
|
Novus Biologicals
NBP3-19756-50ul |
50 μg |
€ 444.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606413
|
Novus Biologicals
NBP3-19756-100ul |
100 μg |
€ 680.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAC3 Monoclonal antibody specifically detects RAC3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| RAC3 | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 5881 | |
| IgG | |
| Affinity purified |
| S01-6I8 | |
| Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| p21-Rac3, ras-related C3 botulinum toxin substrate 3, ras-related C3 botulinum toxin substrate 3 (rho family, small GTP bindingprotein Rac3), rho family, small GTP binding protein Rac3 | |
| A synthetic peptide of human RAC3 (Uniprot # P60763) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title