missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAC3 Recombinant Rabbit Monoclonal Antibody (23GB2175), Invitrogen™

Product Code. 30087861
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30087861 20 μL 20µL
30087886 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30087861 Supplier Invitrogen™ Supplier No. MA553241

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Plasma membrane-associated small GTPase which cycles between an active GTP-bound and inactive GDP-bound state. In active state binds to a variety of effector proteins to regulate cellular responses, such as cell spreading and the formation of actin-based protusions including lamellipodia and membrane ruffles. Promotes cell adhesion and spreading on fibrinogen in a CIB1 and alpha-IIb/beta3 integrin-mediated manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAC3
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2175
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAC3
Gene Accession No. P60763, P60764
Gene Alias OTTMUSP00000004488; p21 Rac3; p21-Rac3; Rac1B; RAC3; RAC3_HUMAN; RAS related C3 botulinum substrate 3; Ras related C3 botulinum toxin substrate 3; Ras-related C3 botulinum toxin substrate 3; Rho family small GTP binding protein Rac3; RP23-84C12.18
Gene Symbols RAC3
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAC3 (100-150AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170758, 5881
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.