missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RACGAP1 Monoclonal Antibody (OTI3A5)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527507
This item is not returnable.
View return policy
Description
RACGAP1 Monoclonal Antibody for Western Blot, IHC (P)
RAC, a small GTPase in the ras family, regulates the formation of membrane ruffles, lamellipodia and filopodia. It is involved in cytoskeletal actin organization and transformation. The p21 activated kinases (PAKs) are direct targets of active Rac and Cdc42 which can induce the assembly of polarized cytoskeletal structures when expressed in fibroblasts. RAC1 and RAC2 are also essential components of NADPH oxidase, the enzyme responsible for generating free radicals.
Specifications
| RACGAP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H0H5 | |
| RACGAP1 | |
| Human recombinant protein fragment corresponding to amino acids 123-238 of RACGAP1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3A5 | |
| Unconjugated | |
| RACGAP1 | |
| AI227039; AI327394; AKT; Band25; CYK4; GAP; gtl11; GTPase; HsCYK-4; ID; ID-GAP; KIAA1478; Male germ cell RacGap; MGC99656; MgcRacGAP; mKIAA1478; PKB; PKB-ALPHA; PRKBA; protein CYK4 homolg; Protein CYK4 homolog; RAC; Rac GTPase activating protein 1; Rac GTPase-activating protein 1; RAC-ALPHA; Racgap1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 29127 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction