missing translation for 'onlineSavingsMsg'
Learn More

RACGAP1 Monoclonal Antibody (OTI4H8), TrueMAB™, OriGene

Product Code. p-200018167
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17545774 30 μL 30µL
17525774 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17545774 Supplier Origene Technologies Supplier No. TA811641S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RACGAP1 Monoclonal antibody specifically detects RACGAP1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen RACGAP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RACGAP1
Gene Accession No. Q9H0H5
Gene Alias AI227039; AI327394; AKT; Band25; CYK4; GAP; gtl11; GTPase; HsCYK-4; ID; ID-GAP; KIAA1478; Male germ cell RacGap; MGC99656; MgcRacGAP; mKIAA1478; PKB; PKB-ALPHA; PRKBA; protein CYK4 homolg; Protein CYK4 homolog; RAC; Rac GTPase activating protein 1; Rac GTPase-activating protein 1; RAC-ALPHA; Racgap1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 123-238 of human RACGAP1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29127
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.