Learn More
RACGAP1 Polyclonal Antibody
RACGAP1 Polyclonal Antibody
Brand:  Invitrogen PA568992
Product Code. 15857403
  • € 530.00 / 100µL
Description
RACGAP1 Polyclonal Antibody for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RACGAP1
Polyclonal
Unconjugated
RACGAP1
AI227039; AI327394; AKT; Band25; CYK4; GAP; gtl11; GTPase; HsCYK-4; ID; ID-GAP; KIAA1478; Male germ cell RacGap; MGC99656; MgcRacGAP; mKIAA1478; PKB; PKB-ALPHA; PRKBA; protein CYK4 homolg; Protein CYK4 homolog; RAC; Rac GTPase activating protein 1; Rac GTPase-activating protein 1; RAC-ALPHA; Racgap1
Rabbit
Affinity Chromatography
RUO
315298
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
0
RACGAP1
synthetic peptide directed towards the C-terminal region of rat RACGAP1.