Learn More
Atrow™ Rack
Click to view available options
Array:
4 x 12
4 x 6
5 x 10
Height (Metric):
100 mm
25 cm
Length (Metric):
103 mm
150 mm
296 mm
Unit Size:
Each
Pack of 30
Specifications
Specifications
| Includes | White rilsan coating |
| Product Type | Rack |
| Array | 4 x 12 |
| Holds | Tubes (48) |
| For Use With (Application) | 16, 18 and 20mm dia. short test tube |
| Length (Metric) | 296 mm |
| Height (Metric) | 100 mm |