Learn More
Atrow™ Rack
Click to view available options
Array:
4 x 12
4 x 6
5 x 10
Height (Metric):
100 mm
25 cm
Length (Metric):
103 mm
150 mm
296 mm
Unit Size:
Each
Pack of 30
Specifications
Specifications
| Array | 5 x 10 |
| Holds | Tubes (50) |
| For Use With (Equipment) | Micro Tubes |
| Length (Metric) | 103 mm |
| Height (Metric) | 25 cm |
| Diameter (Metric) Outer | 11 cm |