Learn More
Atrow™ Rack
Product Code. p-8007836
Click to view available options
Array:
4 x 12
4 x 6
5 x 10
Height (Metric):
100 mm
25 cm
Length (Metric):
103 mm
150 mm
296 mm
Unit Size:
Each
Pack of 30
Product Code. 15990246
Brand: Atrow™ 805108
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Array 5 x 10
Holds Tubes (50)
For Use With (Equipment) Micro Tubes
Length (Metric) 103 mm
Height (Metric) 25 cm
Diameter (Metric) Outer 11 cm