Learn More
Tenak™ Standard Rack for Microtiterplates, for Upright Freezers, 16 mm Shelf Height
Stainlesss Steel Standard Rack for use in upright freezers is designed to hold microtiterplates.
Brand: Tenak™ TE27458
Additional Details : Weight : 7.40000kg
Description
- For use in upright freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of stainless steel
Specifications
Stainless Steel | |
Microtiterplates (115) | |
450 x 135 x 391 mm |
Upright Freezers | |
23 x 5 | |
16 mm |