Learn More
RACK1 Recombinant Polyclonal Antibody (23HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Brand: Invitrogen 711276
Description
This antibody is predicted to react with Monkey, Cat, Sheep Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
RACK1 (receptor for activated C kinase 1) was identified through its binding to various PKC isoforms. Its main function is to recruit PKC and various other proteins to specific locations to form multiprotein complexes, mediating various signal pathways. RACK1 is required for VANGL2 membrane localization and for PTK2/FAK1 phosphorylation and dephosphorylation.
Specifications
| RACK1 | |
| Recombinant Polyclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P63244, P68040 | |
| RACK1 | |
| Peptides corresponding to human RACK1 (aa268-285, aa139-152) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| 23HCLC | |
| Unconjugated | |
| RACK1 | |
| 12-3; activated protein kinase C receptor; AL033335; Cell proliferation-inducing gene 21 protein; CG7111; CG7111-PA; CG7111-PB; CG7111-PC; CG7111-PD; Dmel\CG7111; Dmel_CG7111; dRack1; GB-like; gnb2l1; Gnb2-rs1; guanine nucleotide binding protein (G protein) beta polypeptide 2-like 1; guanine nucleotide binding protein (G protein), beta polypeptide 2 like 1; guanine nucleotide binding protein (G protein), beta polypeptide 2 like 1 sequence 1; guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1; guanine nucleotide binding protein beta polypeptide 2-like 1; guanine nucleotide binding protein related; guanine nucleotide binding protein, beta 2, related sequence 1; guanine nucleotide binding protein, beta polypeptide 2-like 1; guanine nucleotide binding protein, beta-2, related sequence 1; guanine nucleotide-binding protein beta-subunit like protein; guanine nucleotide-binding protein subunit beta-2-like 1; Guanine nucleotide-binding protein subunit beta-2-like 1, N-terminally processed; Guanine nucleotide-binding protein subunit beta-like protein; Guanine nucleotide-binding protein subunit beta-like protein 12.3; H12.3; HLC7; HLC-7; Human lung cancer oncogene 7 protein; i173; lung cancer oncogene 7; OTTHUMP00000223870; OTTHUMP00000223891; OTTHUMP00000223893; OTTHUMP00000223900; OTTHUMP00000223901; OTTHUMP00000223902; OTTHUMP00000223930; OTTHUMP00000223931; p205; PIG21; proliferation-inducing gene 21; protein homologous to chicken B complex protein, guanine nucleotide binding; protein kinase C receptor; RACK; Rack1; rack-1; Rack1-PA; Rack1-PB; Rack1-PC; Rack1-PD; RE74715p; receptor for activated C kinase; receptor for activated C kinase 1; receptor for activated protein kinase C; receptor for activated protein kinase C1; receptor of activated PKC; receptor of activated protein C kinase 1; Receptor of activated protein C kinase 1, N-terminally processed; Receptor of activated protein kinase C; receptor of activated protein kinase C 1; Receptor of activated protein kinase C homolog; similar to mammalian RACK1; Small ribosomal subunit protein RACK1; wu:fb80d08; wu:fk65d12 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10399, 14694 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.