missing translation for 'onlineSavingsMsg'
Learn More

Rad1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-97717-100ul

 View more versions of this product

Product Code. 18659591

  • € 326.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Rad1 Polyclonal antibody specifically detects Rad1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rad1
Polyclonal
Immunocytochemistry/ Immunofluorescence 1:100-1:500
cell cycle checkpoint protein Hrad1, checkpoint control protein HRAD1, DNA repair exonuclease rad1 homolog, DNA repair exonuclease REC1, EC 3.1.11.2, exonuclease homolog RAD1, HRAD1, RAD1 (S. pombe) homolog, RAD1 homolog (S. pombe), Rad1-like DNA damage checkpoint protein, REC1cell cycle checkpoint protein RAD1
Produced in rabbits immunized with E. coli-derived Human Rad1 fragment.
100 μg
Cancer, DNA Repair, Signal Transduction
5810
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Immunofluorescence
Unconjugated
PBS (pH 7.0)
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.