missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD1, Mouse anti-Human, Clone: 1A12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005810-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: MPLLTQQIQDEDDQYSLVASLDNVRNLSTILKAIHFREHATCFATKNGIKVTVENAKCVQANAFIQAGIFQEFKVQEESVTFRINLTVLLSpecifications
| RAD1 | |
| Monoclonal | |
| Unconjugated | |
| RAD1 homolog (S. pombe) | |
| HRAD1/REC1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1A12 | |
| In 1x PBS, pH 7.4 | |
| BC006837 | |
| RAD1 | |
| RAD1 (AAH06837, 1 a.a. ∼ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5810 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction