missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad17 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-172SS
This item is not returnable.
View return policy
Description
Rad17 Polyclonal specifically detects Rad17 in Yeast samples. It is validated for Western Blot.
Specifications
| Rad17 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.01% Sodium Azide | |
| RAD17 | |
| Synthetic peptide made to Schizosaccharomyces pombe RAD17. | |
| Affinity Purified | |
| Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair | |
| 5884 | |
| Yeast | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:500 -1:1000 | |
| CCYC, cell cycle checkpoint protein (RAD17), cell cycle checkpoint protein RAD17, FLJ41520, HRAD17, R24L, RAD1 (S. pombe) homolog, RAD1 homolog, RAD17 homolog (S. pombe), Rad17-like protein, RAD17SP, Rad24, RF-C activator 1 homolog, RF-C/activator 1 homolog | |
| Rabbit | |
| 80 kDa | |
| 0.025 mg | |
| Primary | |
| Reacts with residues 10-23 [STKRSLKKKKIRKI] of the S. pombe RAD17 75 kDA protein. | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction