missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad17 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-172CL1
This item is not returnable.
View return policy
Description
Rad17 Polyclonal antibody specifically detects Rad17 in Yeast samples. It is validated for Western Blot
Specifications
| Rad17 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Yeast | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| CCYC, cell cycle checkpoint protein (RAD17), cell cycle checkpoint protein RAD17, FLJ41520, HRAD17, R24L, RAD1 (S. pombe) homolog, RAD1 homolog, RAD17 homolog (S. pombe), Rad17-like protein, RAD17SP, Rad24, RF-C activator 1 homolog, RF-C/activator 1 homolog | |
| Synthetic peptide made to Schizosaccharomyces pombe RAD17. | |
| 0.1 mL | |
| Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Phospho Specific | |
| 5884 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction