Learn More
RAD17 Mouse, anti-Human, Clone: 1F12D8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68539-1-IG-20UL
 View more versions of this product
Product Code. 18065145
  • € 145.00 / 20µL
Description
The RAD17 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAD17. This antibody recognizes human antigen. The RAD17 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
RAD17
1F12D8
CCYC, FLJ41520, HRAD17, R24L, RAD17, RAD17 homolog (S. pombe), RAD17SP, RAD24, RF C/activator 1 homolog
20 μL
5884
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
500 μg/mL
Mouse
Primary
Human
150 kDa