Learn More
RAD17 Mouse, anti-Human, Clone: 1F12D8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68539-1-IG-20UL
This item is not returnable.
View return policy
Description
The RAD17 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAD17. This antibody recognizes human antigen. The RAD17 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| RAD17 | |
| 1F12D8 | |
| CCYC, FLJ41520, HRAD17, R24L, RAD17, RAD17 homolog (S. pombe), RAD17SP, RAD24, RF C/activator 1 homolog | |
| 20 μL | |
| 5884 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |