missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad17, p Ser645 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-273FR
This item is not returnable.
View return policy
Description
Rad17 Polyclonal antibody specifically detects Rad17 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Rad17 | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry | |
| CCYC, cell cycle checkpoint protein (RAD17), cell cycle checkpoint protein RAD17, FLJ41520, HRAD17, R24L, RAD1 (S. pombe) homolog, RAD1 homolog, RAD17 homolog (S. pombe), Rad17-like protein, RAD17SP, Rad24, RF-C activator 1 homolog, RF-C/activator 1 homolog | |
| Immunogen affinity purified | |
| RUO | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction