missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD18 Antibody (79B1048.1), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56523AF700
This item is not returnable.
View return policy
Description
RAD18 Monoclonal specifically detects RAD18 in Human samples. It is validated for Western Blot.
Specifications
| RAD18 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RAD18 | |
| The antibody was raised against a synthetic peptide corresponding to the amino acids 402-414 of human Rad18. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers, Zinc Finger | |
| 56852 | |
| Human | |
| Purified |
| Western Blot | |
| 79B1048.1 | |
| Western Blot | |
| EC 6.3.2.-, hHR18, hRAD18, postreplication repair protein hRAD18p, Postreplication repair protein RAD18, RAD18 homolog (S. cerevisiae), RAD18, S. cerevisiae, homolog, RING finger protein 73, RNF73E3 ubiquitin-protein ligase RAD18 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| In HL60, a 54 kDa band should be detected. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction