missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad21 Antibody (CM110-2C10), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-386AF700
This item is not returnable.
View return policy
Description
Rad21 Monoclonal specifically detects Rad21 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Rad21 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| O60216 | |
| RAD21 | |
| An N-terminal fusion protein of the C-terminal fragment of Scc1 subunit human cohesin (amino acids 544-631 of hRad21). | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair | |
| 5885 | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| CM110-2C10 | |
| Western Blot | |
| double-strand-break repair protein rad21 homolog, hHR21FLJ25655, HR21, KIAA0078RAD21 (S. pombe) homolog, MCD1, Nuclear matrix protein 1, NXP-1, NXP1HRAD21, protein involved in DNA double-strand break repair, RAD21 homolog (S. pombe), SCC1 homolog, SCC1FLJ40596 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-386 is specific for both the nuclear hRad21 protein and the cleaved cytoplasmic hRad21 protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction