Learn More
Rad21 Antibody (CM110-2C10), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-386AF750
Product Code. 18040514
  • € 555.00 / 0.10mL
Description
Rad21 Monoclonal specifically detects Rad21 in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rad21
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
RAD21
An N-terminal fusion protein of the C-terminal fragment of Scc1 subunit human cohesin (amino acids 544-631 of hRad21).
Primary
NB100-386 is specific for both the nuclear hRad21 protein and the cleaved cytoplasmic hRad21 protein.
Store at 4°C in the dark.
Western Blot
CM110-2C10
Western Blot
double-strand-break repair protein rad21 homolog, hHR21FLJ25655, HR21, KIAA0078RAD21 (S. pombe) homolog, MCD1, Nuclear matrix protein 1, NXP-1, NXP1HRAD21, protein involved in DNA double-strand break repair, RAD21 homolog (S. pombe), SCC1 homolog, SCC1FLJ40596
Mouse
0.1 mL
5885