missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD21 Monoclonal Antibody (OTI2A12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811221
This item is not returnable.
View return policy
Description
The cohesin complex plays an important role in tethering sister chromatids during the prophase through anaphase stages of mitosis, making certain that genomic information is replicated accurately. As the cellular division process continues, separase destroys the cohesin complex by means of cleavaging its Scc1 subunit, allowing the chromatids to separate and divide with the cell. During apoptosis the C-terminal portion of hRad21 is cleaved and translocated to the cytoplasm. This cleavage and translocation may act as a nuclear signal that promotes and accelerates subsequent events of apoptosis.Specifications
| RAD21 | |
| Monoclonal | |
| Unconjugated | |
| RAD21 | |
| CDLS4, HR21, HRAD21, MCD1, NXP1, SCC1, hHR21 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5885 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A12 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O60216 | |
| RAD21 | |
| Human recombit protein fragment corresponding to amino acids 193-458 of human RAD21 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction