missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD21 Monoclonal Antibody (OTI6C10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811222S
This item is not returnable.
View return policy
Description
RAD21 Monoclonal antibody specifically detects RAD21 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| RAD21 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O60216 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5885 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6C10 | |
| Unconjugated | |
| Rad21 | |
| 64-kDa carboxy-terminal product; 64-kDa C-terminal product; 65-kDa carboxy-terminal product; CDLS4; double-strand-break repair protein rad21 homolog; double-strand-break repair protein rad21-like protein; FLJ25655; FLJ40596; h NXP-1; hHR21; HR21; HRAD21; KIAA0078; KIAA0078RAD21; kleisin; MCD1; mHR21; mKIAA0078; nuclear matrix protein 1; NXP1; NXP-1; Pokeweed agglutinin-binding protein 29; protein involved in DNA double-strand break repair; PW29; Rad21; RAD21 cohesin complex component; RAD21 homolog; RAD21 homolog (S. pombe); SCC1; SCC1 homolog; sister chromatid cohesion 1 | |
| Human recombinant protein fragment corresponding to amino acids 193-458 of human RAD21 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction