missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAD21 Recombinant Rabbit Monoclonal Antibody (BLR052F)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544297
This item is not returnable.
View return policy
Description
RAD21 Recombinant Monoclonal Antibody for Western Blot, Flow
This gene encodes a nuclear protein that regulates transcriptional elongation and pre-mRNA splicing. The encoded protein interacts with the hyperphosphorylated C-terminal domain of RNA polymerase II via multiple FF domains, and with the pre-mRNA splicing factor SF1 via a WW domain. Alternative splicing results in multiple transcripts variants encoding different isoforms.
Specifications
| RAD21 | |
| Recombinant Monoclonal | |
| 40 μg/mL | |
| BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6 | |
| O60216, Q61550 | |
| Rad21 | |
| Peptide representing a region between residue 581 and the C-terminus (residue 631) of human Double-strand-break repair protein Rad21 homolog. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Western Blot | |
| BLR052F | |
| Unconjugated | |
| Rad21 | |
| 64-kDa carboxy-terminal product; 64-kDa C-terminal product; 65-kDa carboxy-terminal product; CDLS4; double-strand-break repair protein rad21 homolog; double-strand-break repair protein rad21-like protein; FLJ25655; FLJ40596; h NXP-1; hHR21; HR21; HRAD21; KIAA0078; KIAA0078RAD21; kleisin; MCD1; mHR21; mKIAA0078; nuclear matrix protein 1; NXP1; NXP-1; Pokeweed agglutinin-binding protein 29; protein involved in DNA double-strand break repair; PW29; Rad21; RAD21 cohesin complex component; RAD21 homolog; RAD21 homolog (S. pombe); SCC1; SCC1 homolog; sister chromatid cohesion 1 | |
| Rabbit | |
| purified | |
| RUO | |
| 19357, 5885 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction