missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD23B Monoclonal Antibody (OTI13A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804864
This item is not returnable.
View return policy
Description
The protein encoded by this gene is one of two human homologs of Saccharomyces cerevisiae Rad23, a protein involved in the nucleotide excision repair (NER). This protein was found to be a component of the protein complex that specifically complements the NER defect of xeroderma pigmentosum group C (XP-c) cell extracts in vitro. This protein was also shown to interact with, and elevate the nucleotide excision activity of 3-methyladenine-DNA glycosylase (MPG), which suggested a role in DNA damage recognition in base excision repair. This protein contains an N-terminal ubiquitin-like domain, which was reported to interact with 26S proteasome, and thus this protein may be involved in the ubiquitin mediated proteolytic pathway in cells.Specifications
| RAD23B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P54727 | |
| RAD23B | |
| Human recombit protein fragment corresponding to amino acids 1-253 of human RAD23B produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI13A3 | |
| Unconjugated | |
| RAD23B | |
| HHR23B, HR23B, P58 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5887 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction