missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad50 Antibody (JE60-04), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32890
This item is not returnable.
View return policy
Description
Rad50 Monoclonal antibody specifically detects Rad50 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| Rad50 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| DNA repair protein RAD50, EC 3.6, EC 3.6.1.15, EC 3.6.3, EC 3.6.3.27, hRAD50, NBSLD, RAD50 (S. cerevisiae) homolog, RAD50 homolog (S. cerevisiae), RAD502, RAD50-2 | |
| Recombinant protein within Human Rad50 aa 1,162-1,312/ 1,312. (Uniprot: Q92878) | |
| 100 μL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining | |
| 10111 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE60-04 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction