missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAD50 Monoclonal Antibody (13B3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA123269
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa, HeLa nuclear extract.
Rad50-13B3 recognizes human Rad50, a 153a protein involved in DNA double strand break repair. Rad50 is associated with Mre11 and p95 (Nibrin) to form a multiprotein complex involved in the double strand break repair process. Upon DNA damage, Rad50 and Mre11 redistribute and are co-localized within the nucleus of damaged cells.
Specifications
| RAD50 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P70388, Q92878, Q9JIL8 | |
| RAD50 | |
| Amino acids 1-425 of Rad50 expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG1 |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Proximity Ligation Assay, Immunocytochemistry | |
| 13B3 | |
| Unconjugated | |
| RAD50 | |
| 153 kDa protein; DNA repair protein RAD50; hRad50; hsRAD50; I79_004319; mRad50; Mrell; MRX complex DNA-binding subunit; N0872; NBSLD; Rad50; RAD50 2; RAD50 double strand break repair protein; RAD50 homolog (S. cerevisiae); RAD50 homolog, double strand break repair protein; RAD50 PEN; RAD502; RAD50-2; RAD50-3; Rad50l; YNL250W | |
| Mouse | |
| Protein G | |
| RUO | |
| 10111, 19360, 64012 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction