missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD51AP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11255-1-AP-150UL
This item is not returnable.
View return policy
Description
May participate in a common DNA damage response pathway associated with the activation of homologous recombination and double-strand break repair. Functionally cooperates with PALB2 in promoting of D-loop formation by RAD51. Binds to single and double stranded DNA, and is capable of aggregating DNA. Also binds RNA.Specifications
| RAD51AP1 | |
| Polyclonal | |
| Unconjugated | |
| RAD51AP1 | |
| PIR51, RAD51 associated protein 1, RAD51 interacting protein, RAD51AP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10635, 19362, 689055 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8C551, Q96B01 | |
| RAD51AP1 | |
| RAD51AP1 Fusion Protein Ag1781 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction