missing translation for 'onlineSavingsMsg'
Learn More

Rad51C Antibody (Rad51C 2H11/6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

€ 190.00 - € 351.00

Specifications

Antigen Rad51C
Clone Rad51C 2H11/6
Concentration 1 mg/mL
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18699448
View Documents
Novus Biologicals
NBP2-50263
0.1 mL
€ 351.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18657508
View Documents
Novus Biologicals
NBP2-50263-0.025ml
0.025 mL
€ 190.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Rad51C Monoclonal antibody specifically detects Rad51C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications

Specifications

Rad51C
1 mg/mL
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination
PBS
5889
IgG1
Protein A purified
Rad51C 2H11/6
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Monoclonal
Purified
RUO
Human
BROVCA3, DNA repair protein RAD51 homolog 3, FANCO, MGC104277, R51H3, RAD51 homolog C, RAD51 homolog C (S. cerevisiae), RAD51L2RAD51 (S. cerevisiae) homolog C, RAD51-like protein 2, yeast RAD51 homolog 3
Rad51C 2H11/6 was raised against His-tagged human Rad51C, overexpressed in e.coli and purified on a talon affinity column under denaturing conditions, followed by gel purification on a SDS-Page
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.