missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad51D Antibody (Rad51D 5A8/4), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50264AF750
This item is not returnable.
View return policy
Description
Rad51D Monoclonal specifically detects Rad51D in Human, Hamster samples. It is validated for Western Blot, ELISA.
Specifications
| Rad51D | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| RAD51D | |
| Human recombinant denatured protein. | |
| 0.1 mL | |
| Primary | |
| Human, Hamster | |
| IgG1 |
| Western Blot, ELISA | |
| Rad51D 5A8/4 | |
| Western Blot, ELISA | |
| DNA repair protein RAD51 homolog 4, HsTRAD, R51H3Trad, RAD51 homolog D, RAD51DRAD51 (S. cerevisiae)-like 3, RAD51-like 3 (S. cerevisiae), RAD51-like protein 3, recombination repair protein, TRAD | |
| Mouse | |
| Protein A purified | |
| Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination | |
| 5892.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction