missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad51L1 Antibody (OTI7H4), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73782AF647
This item is not returnable.
View return policy
Description
Rad51L1 Monoclonal specifically detects Rad51L1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| Rad51L1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RAD51B | |
| Recombinant protein fragment corresponding to amino acids 119-346 of human Rad51L1 (NP_598193) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI7H4 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| DNA repair protein RAD51 homolog 2, MGC34245, R51H2REC2hREC2, RAD51 (S. cerevisiae)-like 1, RAD51 homolog B, RAD51B, RAD51-like 1 (S. cerevisiae), RAD51-like protein 1, RecA-like protein, recombination repair protein | |
| Mouse | |
| Affinity Purified | |
| Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination | |
| 5890.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction