missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAD52 Monoclonal Antibody (3E11B2E8C11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531889
This item is not returnable.
View return policy
Description
MA5-31889 has been tested in indirect ELISA.
DNA double-strand breaks can be generated by exposure to ionizing radiation or through endogenous cellular reactions. Double-strand breaks are generally thought to be repaired via one of two distinct mechanisms: homologous recombination (HR) or non-homologous end joining (NHEJ). Homologous recombination requires the activity of the members of the RAD52 epistasis group, which includeRAD50, RAD51, RAD52, RAD54, RDH54/TID1, RAD55, RAD57, RAD59, MRE11, and XRS2. RAD52 can catalyze strand annealing reactions, and can form oligomers that attach to ends of single-stranded DNA.
Specifications
| RAD52 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P43351 | |
| RAD52 | |
| Purified recombinant fragment of human RAD52 (AA: 269-418) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 3E11B2E8C11 | |
| Unconjugated | |
| RAD52 | |
| DNA repair protein RAD52 homolog; RAD52; RAD52 homolog (S. cerevisiae); RAD52 homolog, DNA repair protein; Rad52yh; recombination protein RAD52; rhabdomyosarcoma antigen MU-RMS-40.23 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5893 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction