missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAD54L Polyclonal Antibody
GREENER_CHOICE

Product Code. 16667814
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16667814 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16667814 Supplier Invitrogen™ Supplier No. PA5101338

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RAD54.

The protein encoded by this gene belongs to the DEAD-like helicase superfamily, and shares similarity with Saccharomyces cerevisiae Rad54, a protein known to be involved in the homologous recombination and repair of DNA. This protein has been shown to play a role in homologous recombination related repair of DNA double-strand breaks. The binding of this protein to double-strand DNA induces a DNA topological change, which is thought to facilitate homologous DNA paring, and stimulate DNA recombination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD54L
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAD54L
Gene Accession No. P70270, Q92698
Gene Alias ATR2; DNA repair and recombination protein RAD54-like; h hRAD54; hHR54; HR54; hRAD54; MGC2094; mHR54; mRAD54; MRXHF1; Rad54; RAD54 homolog; RAD54 like (S. cerevisiae); RAD54A; RAD54L; RAD54-like (S. cerevisiae); RP5-875J14.1; SFM1; SHS; XH2; XNP; ZNF-HX
Gene Symbols RAD54L
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAD54L(Accession Q92698), corresponding to amino acid residues T675-D725.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19366, 8438
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.