missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD54L2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93717-0.1ml
This item is not returnable.
View return policy
Description
RAD54L2 Polyclonal antibody specifically detects RAD54L2 in Human samples. It is validated for Western Blot
Specifications
| RAD54L2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Androgen receptor-interacting protein 4, ARIP4, EC 3.6.1, EC 3.6.4.12, FLJ21396, FLJ22400, helicase ARIP4, KIAA0809HSPC325, RAD54-like 2 (S. cerevisiae), RAD54-like protein 2, SRISNF2L | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1178-1467 of human RAD54L2 (NP_055921.2). ELQQYADVAAARESRQSSPSTNAALPGPPAQLMDSSAVPGTALGTEPRLGGHCLNSSLLVTGQPCGDRHPVLDLRGHKRKLATPPAAQESSRRRSRKGHLPAPVQPYEHGYPVSGGFAMPPVSLNHNLTTPFTSQAGENSLFMGSTPSYYQLSNLLADARLVFPVTTDPLVPAGPVSSSSTATSVTASNPSFMLNPSVPGILPSYSLPFSQPLLSEPRMFAPFPSPVLPSNLSRGMSIYPGYMSPHAGYPAGGLLRSQVPPFDSHEVAEVGFSSNDDEDKDDDVIEVTGK | |
| 0.1 mL | |
| Chromatin Modifiers, Signal Transduction | |
| 23132 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction