Learn More
Invitrogen™ Radixin Recombinant Rabbit Monoclonal Antibody (24GB1420)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557675
This item is not returnable.
View return policy
Description
Radixin Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Radixin is a cytoskeletal protein that may be important in linking actin to the plasma membrane. It is highly similar in sequence to both ezrin and moesin. The radixin gene has been localized by fluorescence in situ hybridization to 11q23. A truncated version representing a pseudogene (RDXP2) was assigned to Xp21.3. Another pseudogene that seemed to lack introns (RDXP1) was mapped to 11p by Southern and PCR analyses. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, May 2012].
Specifications
| Radixin | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Rdx | |
| AA516625; DFNB24; ESP10; radixin; radixin isoform; Rdx | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19684, 315655, 5962 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1420 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P26043, P35241 | |
| Rdx | |
| A synthesized peptide derived from human Radixin (aa 350-430). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |