Learn More
Rae-1 Antibody (199205), Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB17581B
This item is not returnable.
View return policy
Description
Rae-1 Monoclonal antibody specifically detects Rae-1 in Mouse samples. It is validated for Flow Cytometry, Blocking Assay, CyTOF
Specifications
| Rae-1 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8480 | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry, Blocking Assay, CyTOF | |
| 199205 | |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| dJ481F12.3,dJ800J21.1,Gle2,MIG14,Mnrp41,mRNA export factor RAE1,MRNP41,RAE1 | |
| BaF3 mouse pro-B cell line transfected with Rae-1 beta (Accession # NP_033043) and Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta Leu29-Ser227 (Accession # Q9JI58) | |
| 0.5 mL | |
| Primary | |
| Mouse | |
| Purified |